Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-15
Entrez Gene ID
183195
Gene Name
Serpentine receptor class gamma-15
Gene Aliases
CELE_C34C6.1
Chromosome Location
Chr.II: 8708689 - 8710417 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063450.3 NP_495851.2 5 - 6 841 80

View Full Product Details