Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rer-1
Entrez Gene ID
174410
Gene Name
Protein RER1 homolog
Gene Aliases
CELE_F46C5.8
Chromosome Location
Chr.II: 8811370 - 8812634 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381540.1 NP_001366730.1 1 - 2 65 70

View Full Product Details