Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-46
Entrez Gene ID
174415
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_T14D7.2
Chromosome Location
Chr.II: 8848843 - 8852096 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063485.6 NP_495886.2 4 - 5 1268 70

View Full Product Details