Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ooc-5
Entrez Gene ID
174433
Gene Name
protein-coding
Gene Aliases
CELE_C18E9.11
Chromosome Location
Chr.II: 8993160 - 8994535 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063515.3 NP_495916.1 3 - 4 396 90
NM_063516.5 NP_495917.1 3 - 4 369 90

View Full Product Details