Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dep-1
Entrez Gene ID
174437
Gene Name
protein-coding
Gene Aliases
CELE_F44G4.8
Chromosome Location
Chr.II: 9005796 - 9020815 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267263.3 NP_001254192.1 8 - 9 1746 98

View Full Product Details