Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clh-5
Entrez Gene ID
174449
Gene Name
Chloride channel protein
Gene Aliases
CELE_C07H4.2
Chromosome Location
Chr.II: 9094710 - 9098605 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063539.6 NP_495940.1 1 - 2 107 112
GenBank mRNA AB035090.2 - 1 - 2 105 112

View Full Product Details