Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rhy-1
Entrez Gene ID
174455
Gene Name
Regulator of hypoxia-inducible factor 1
Gene Aliases
CELE_W07A12.7
Chromosome Location
Chr.II: 9161379 - 9163448 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063553.5 NP_495954.1 1 - 2 192 69

View Full Product Details