Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eat-3
Entrez Gene ID
174476
Gene Name
Dynamin-type G domain-containing protein
Gene Aliases
CELE_D2013.5
Chromosome Location
Chr.II: 9324866 - 9329665 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063585.9 NP_495986.3 8 - 9 2853 84

View Full Product Details