Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
scp-1
Entrez Gene ID
174479
Gene Name
protein-coding
Gene Aliases
CELE_D2013.8
Chromosome Location
Chr.II: 9334012 - 9338902 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026880.5 NP_001022051.1 6 - 7 1838 93
NM_001383916.1 6 - 7 1830 93

View Full Product Details