Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-48
Entrez Gene ID
174486
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_C06C3.6
Chromosome Location
Chr.II: 9378531 - 9384995 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063598.7 NP_495999.2 7 - 8 1826 110

View Full Product Details