Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-1
Entrez Gene ID
191639
Gene Name
Receptor-type guanylate cyclase gcy-1
Gene Aliases
CELE_AH6.1
Chromosome Location
Chr.II: 9512490 - 9516936 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063638.1 NP_496039.1 9 - 10 1354 76

View Full Product Details