Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sra-2
Entrez Gene ID
191773
Gene Name
Serpentine receptor class alpha-2
Gene Aliases
CELE_AH6.6
Chromosome Location
Chr.II: 9528780 - 9529862 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063643.1 NP_496044.1 2 - 3 835 176

View Full Product Details