Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sra-9
Entrez Gene ID
191780
Gene Name
Serpentine receptor class alpha-9
Gene Aliases
CELE_AH6.14
Chromosome Location
Chr.II: 9532311 - 9533974 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063645.5 NP_496046.1 1 - 2 650 171

View Full Product Details