Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sra-3
Entrez Gene ID
191774
Gene Name
Serpentine receptor class alpha-3
Gene Aliases
CELE_AH6.7
Chromosome Location
Chr.II: 9539274 - 9540823 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063647.4 NP_496048.1 1 - 2 648 141

View Full Product Details