Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hlh-6
Entrez Gene ID
188539
Gene Name
Helix-loop-helix protein 6
Gene Aliases
CELE_T15H9.3
Chromosome Location
Chr.II: 9613463 - 9614828 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063669.2 NP_496070.1 1 - 2 225 146

View Full Product Details