Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arc-1
Entrez Gene ID
174525
Gene Name
E3 ubiquitin-protein ligase arc-1
Gene Aliases
CELE_ZK1320.6
Chromosome Location
Chr.II: 9674978 - 9677220 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063688.8 NP_496089.3 4 - 5 1459 91

View Full Product Details