Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-2
Entrez Gene ID
174532
Gene Name
ZP domain-containing protein
Gene Aliases
CELE_Y53C12B.6
Chromosome Location
Chr.II: 9721758 - 9724673 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063696.5 NP_496097.1 5 - 6 988 125

View Full Product Details