Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-13A3
Entrez Gene ID
188359
Gene Name
Putative cytochrome P450 CYP13A3
Gene Aliases
CELE_T10B9.5
Chromosome Location
Chr.II: 9788797 - 9793297 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063709.3 NP_496110.1 2 - 3 533 89

View Full Product Details