Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-4
Entrez Gene ID
174544
Gene Name
Homeobox protein unc-4
Gene Aliases
CELE_F26C11.2
Chromosome Location
Chr.II: 9897751 - 9900376 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063737.2 NP_496138.1 2 - 3 409 81

View Full Product Details