Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ufd-3
Entrez Gene ID
3565222
Gene Name
protein-coding
Gene Aliases
CELE_C05C10.6
Chromosome Location
Chr.II: 9939116 - 9944107 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063745.3 NP_496146.1 6 - 7 1094 91
NM_063746.5 NP_496147.1 2 - 3 367 91

View Full Product Details