Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lec-3
Entrez Gene ID
174557
Gene Name
protein-coding
Gene Aliases
CELE_ZK892.1
Chromosome Location
Chr.II: 9985007 - 10000351 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027360.5 NP_001022531.1 3 - 4 321 88
NM_001027361.4 NP_001022532.1 2 - 3 439 88
NM_001027362.7 NP_001022533.1 2 - 3 409 88
NM_001027363.4 NP_001022534.1 1 - 2 676 88
NM_001276739.3 NP_001263668.1 3 - 4 233 88
NM_063758.8 NP_496159.1 3 - 4 268 88
GenBank mRNA AB038504.1 - 2 - 3 175 88

View Full Product Details