Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lact-2
Entrez Gene ID
174570
Gene Name
Beta-lactamase domain-containing protein 2
Gene Aliases
CELE_ZK945.1
Chromosome Location
Chr.II: 10094873 - 10098785 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063775.5 NP_496176.1 1 - 2 104 55

View Full Product Details