Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
daf-19
Entrez Gene ID
174577
Gene Name
protein-coding
Gene Aliases
CELE_F33H1.1
Chromosome Location
Chr.II: 10157520 - 10169354 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026968.4 NP_001022139.1 2 - 3 283 115
NM_001026969.4 NP_001022140.1 2 - 3 282 115
NM_001267307.4 NP_001254236.1 2 - 3 295 115
GenBank mRNA AF226156.1 - 2 - 3 305 115
AF233652.1 - 2 - 3 305 115

View Full Product Details