Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-1
Entrez Gene ID
191794
Gene Name
Serpentine receptor class delta-1
Gene Aliases
CELE_F33H1.5
Chromosome Location
Chr.II: 10180199 - 10182734 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063794.1 NP_496195.1 3 - 4 491 96

View Full Product Details