Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-4
Entrez Gene ID
191642
Gene Name
Receptor-type guanylate cyclase gcy-4
Gene Aliases
CELE_ZK970.5
Chromosome Location
Chr.II: 10304139 - 10308826 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063817.2 NP_496218.2 7 - 8 1203 135

View Full Product Details