Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-5
Entrez Gene ID
191643
Gene Name
Receptor-type guanylate cyclase gcy-5
Gene Aliases
CELE_ZK970.6
Chromosome Location
Chr.II: 10310400 - 10316219 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063818.1 NP_496219.1 7 - 8 1197 75

View Full Product Details