Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pro-2
Entrez Gene ID
174598
Gene Name
Nucleolar complex protein 2 homolog
Gene Aliases
CELE_C07E3.2
Chromosome Location
Chr.II: 10339692 - 10345112 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063823.5 NP_496224.2 3 - 4 776 107
GenBank mRNA EU068465.1 - 3 - 4 775 107

View Full Product Details