Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
taf-13
Entrez Gene ID
192052
Gene Name
TAF (TBP-associated transcription factor) family
Gene Aliases
CELE_C14A4.10
Chromosome Location
Chr.II: 10605030 - 10605612 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063888.5 NP_496289.2 1 - 2 140 94

View Full Product Details