Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-9
Entrez Gene ID
174640
Gene Name
ZP domain-containing protein
Gene Aliases
CELE_M28.1
Chromosome Location
Chr.II: 10622265 - 10625946 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063892.6 NP_496293.2 3 - 4 713 136

View Full Product Details