Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- vps-11
- Entrez Gene ID
-
174661
- Gene Name
- Vacuolar protein sorting-associated protein 11 homolog
- Gene Aliases
-
CELE_R06F6.2
- Chromosome Location
-
Chr.II: 10788106 - 10793493 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063921.7 |
NP_496322.2 |
2 - 3 |
888 |
90 |
View Full Product Details