Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npp-19
Entrez Gene ID
174663
Gene Name
protein-coding
Gene Aliases
CELE_R06F6.5
Chromosome Location
Chr.II: 10796646 - 10798479 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063923.6 NP_496324.1 1 - 2 75 76
NM_063924.6 NP_496325.1 1 - 2 75 76

View Full Product Details