Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ech-4
Entrez Gene ID
174665
Gene Name
ACB domain-containing protein
Gene Aliases
CELE_R06F6.9
Chromosome Location
Chr.II: 10821031 - 10822449 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381584.1 NP_001366707.1 2 - 3 654 89

View Full Product Details