Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ram-2
Entrez Gene ID
174687
Gene Name
Col_cuticle_N domain-containing protein
Gene Aliases
CELE_F38A3.2
Chromosome Location
Chr.II: 11015589 - 11016881 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063961.5 NP_496362.1 1 - 2 141 97

View Full Product Details