Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mat-2
Entrez Gene ID
174698
Gene Name
Anaphase-promoting complex subunit 1
Gene Aliases
CELE_W10C6.1
Chromosome Location
Chr.II: 11112719 - 11121107 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063982.4 NP_496383.2 8 - 9 3843 65

Gene Symbol
apc-17
Entrez Gene ID
182479
Gene Name
Anaphase Promoting Complex; see also mat
Gene Aliases
CELE_C09H10.7
Chromosome Location
Chr.II: 11108128 - 11120776 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063980.3 NP_496381.1 11 - 12 4844 65

View Full Product Details