Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqm-1
Entrez Gene ID
174705
Gene Name
C2H2-type domain-containing protein
Gene Aliases
CELE_F40F8.7
Chromosome Location
Chr.II: 11150299 - 11152604 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063990.5 NP_496391.1 2 - 3 477 146
GenBank mRNA AF008590.1 - 2 - 3 437 146

View Full Product Details