Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clh-2
Entrez Gene ID
174738
Gene Name
protein-coding
Gene Aliases
CELE_B0491.8
Chromosome Location
Chr.II: 11351899 - 11370320 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026729.4 NP_001021900.1 15 - 16 2439 85
NM_001377875.3 NP_001364585.1 15 - 16 2517 85
NM_001383953.1 14 - 15 2327 85
GenBank mRNA AF173171.1 - 14 - 15 2349 85

View Full Product Details