The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001027219.6 | NP_001022390.1 | 6 - 7 | 755 | 68 |
NM_001027220.5 | NP_001022391.1 | 2 - 3 | 155 | 68 |