Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttr-18
Entrez Gene ID
174757
Gene Name
TransThyretin-Related family domain
Gene Aliases
CELE_B0334.1
Chromosome Location
Chr.II: 11485508 - 11486748 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064052.8 NP_496453.1 1 - 2 155 99

View Full Product Details