Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rad-23
Entrez Gene ID
174785
Gene Name
Uncharacterized protein
Gene Aliases
CELE_ZK20.3
Chromosome Location
Chr.II: 11651257 - 11652729 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064087.8 NP_496488.2 1 - 2 68 90

View Full Product Details