Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
trr-1
Entrez Gene ID
174788
Gene Name
protein-coding
Gene Aliases
CELE_C47D12.1
Chromosome Location
Chr.II: 11661569 - 11675927 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001377877.2 NP_001364647.1 1 - 2 133 121
NM_001377878.3 NP_001364646.1 1 - 2 133 121
NM_001377879.1 NP_001364592.1 1 - 2 204 121

View Full Product Details