Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dna-2
Entrez Gene ID
174809
Gene Name
yeast DNA helicase/endonuclease family
Gene Aliases
CELE_F43G6.1
Chromosome Location
Chr.II: 11778430 - 11782396 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064115.4 NP_496516.1 6 - 7 1148 94

View Full Product Details