Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cnt-1
Entrez Gene ID
174848
Gene Name
protein-coding
Gene Aliases
CELE_Y17G7B.15
Chromosome Location
Chr.II: 12072067 - 12088868 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383965.1 5 - 6 619 67

View Full Product Details