Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cnt-1
Entrez Gene ID
174848
Gene Name
protein-coding
Gene Aliases
CELE_Y17G7B.15
Chromosome Location
Chr.II: 12072067 - 12088868 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027242.3 NP_001022413.1 4 - 5 1620 111
NM_001348640.3 NP_001335541.1 1 - 2 360 111
NM_001381624.1 NP_001368630.1 4 - 5 1116 111
NM_001383965.1 9 - 10 1876 111

View Full Product Details