Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-87
Entrez Gene ID
174861
Gene Name
protein-coding
Gene Aliases
CELE_Y57A10A.18
Chromosome Location
Chr.II: 12209095 - 12225086 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267406.4 NP_001254335.1 3 - 4 253 56
NM_001267407.4 NP_001254336.1 3 - 4 411 56
NM_001381625.2 NP_001367160.1 4 - 5 494 56
NM_001381626.2 NP_001368162.1 3 - 4 408 56
NM_064193.4 NP_496594.2 4 - 5 519 56

View Full Product Details