Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-87
Entrez Gene ID
174861
Gene Name
protein-coding
Gene Aliases
CELE_Y57A10A.18
Chromosome Location
Chr.II: 12209095 - 12225086 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267406.4 NP_001254335.1 7 - 8 1320 76
NM_001267407.4 NP_001254336.1 7 - 8 1478 76
NM_001381625.2 NP_001367160.1 8 - 9 1561 76
NM_001381626.2 NP_001368162.1 7 - 8 1475 76
NM_064193.4 NP_496594.2 8 - 9 1586 76

View Full Product Details