Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rsr-2
Entrez Gene ID
174862
Gene Name
cwf21 domain-containing protein
Gene Aliases
CELE_Y57A10A.19
Chromosome Location
Chr.II: 12225356 - 12228209 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383969.1 2 - 3 1075 59

View Full Product Details