Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-27
Entrez Gene ID
190348
Gene Name
Serpentine receptor class epsilon-27
Gene Aliases
CELE_Y57A10C.3
Chromosome Location
Chr.II: 12415527 - 12417614 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064235.2 NP_496636.1 4 - 5 964 64

View Full Product Details