Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-16
Entrez Gene ID
184513
Gene Name
Serpentine receptor class gamma-16
Gene Aliases
CELE_F15A4.4
Chromosome Location
Chr.II: 12467133 - 12469369 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064255.3 NP_496656.2 2 - 3 1010 113

View Full Product Details