Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-4
Entrez Gene ID
189654
Gene Name
C-type LECtin
Gene Aliases
CELE_Y38E10A.5
Chromosome Location
Chr.II: 12578331 - 12581113 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064287.4 NP_496688.1 1 - 2 267 70

View Full Product Details