Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tbc-17
Entrez Gene ID
174900
Gene Name
protein-coding
Gene Aliases
CELE_Y46G5A.1
Chromosome Location
Chr.II: 12694410 - 12726980 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027263.7 NP_001022434.2 12 - 13 2674 70
NM_001027264.7 NP_001022435.2 12 - 13 2676 70
NM_001267427.4 NP_001254356.1 6 - 7 1780 70

View Full Product Details